Ion chromatography and water purification system, PURELAB® Flex 5
Fournisseur: ELGA LabWater
PF5XXXXM1
ELGAPF5XXXXM1EA 0 EUR InStock
ELGAPF5XXXXM1
Ion chromatography and water purification system, PURELAB® Flex 5
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
This all-in-one system is designed to couple directly with the Metrohm 941 Eluent Production Module. It has an integrated recirculation chamber that ensures a supply of ultrapure water (Type I) to Metrohm ion chromatographs that is guaranteed to be fresh.
  • Complete automation: No need to monitor eluent concentration make-up, no need to refill or to set reminders to refill the water
  • Integral built-in reservoir for pressureless ultrapure water supply
  • Closed system avoids eluent contamination due to water handling and exposure
  • Monitor water purity via USB datalog ensuring analytical performance during test batch
  • Prevent column clogging and increases column life: UV (185/254 nm) ensures bacteria and organics control
  • Stable retention times: no dissolved CO₂ in water prevents pH change
  • Enhanced detection limits through improved stability and purity
  • Simultaneous multi-usage: eluent generation, system rinsing and general laboratory work
  • Reduces eluent preparation to minimum compared to using bottled water
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité