Ultrasonic baths, Elmasonic x-tra ST series
Fournisseur: ELMA SCHMIDBAUER
1076429 1076431 1076438 1076439 1076442
ELMA1076429EA 3570 EUR InStock
ELMA1076429 ELMA1076431 ELMA1076438 ELMA1076439 ELMA1076442
Ultrasonic baths, Elmasonic x-tra ST series
Bains à ultrasons
Elmasonic xtra ST floor-mounted ultrasonic devices are designed for use in a production environment, workshops and servicing. The wide range of features makes it easy and effective to work with these devices.
  • Mobile single-tank devices
  • Switchable Dynamic, Pulse and Sweep-functions for more ultrasonic performance
  • Long-lasting, robust devices with tanks made of special stainless steel
  • Easy and clear operation, protected against spraying water
  • Short heating times due to the high heating capacity with temperature regulation (30 to 80 °C)
Multi-frequency at 25 kHz: For rough part cleaning and removing stubborn contaminants like lapping and polishing paste.
Multi-frequency at 45 kHz: For cleaning sensitive parts and surfaces from contaminants like oils, drilling emulsions and greases. Also ideal for hard metal and glass surfaces in industrial and artisan environments as well as jewellery manufactures.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité