Redox Sensor for Standard Applications, CPS12
Fournisseur: Endress+Hauser
CPS12-0PA2GSA
ENDRCPS12-0PA2GSAEA 421 EUR InStock
ENDRCPS12-0PA2GSA
Redox Sensor for Standard Applications, CPS12
Électrodes Électrodes redox
Glass ORP sensor for standard applications.
  • Robust electrode with long poison diffusion path
  • Low maintenance thanks to large, dirt-repellent PTFE ring diaphragm
  • Suitable for harsh applications: process glass for highly alkaline media and pressure-stable up to 16 bar (232 psi)
  • Available with various measuring elements for application in oxidizing or reducing media
  • Memosens technology offers maximum process safety through contactless inductive signal transmission
The Orbisint CPS12 is the all-rounder for ORP measurement in process and environmental applications. Designed for low maintenance and a long operating life.
Material: Platinum measuring surface
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité