Cartridge filter, Guardian™ HPX
Fournisseur: ENTEGRIS INC
GHPV01P01
ENTEGHPV01P01EA 0 EUR InStock
ENTEGHPV01P01
Cartridge filter, Guardian™ HPX
Filtres Cartouches filtrantes
This cartridge filter with high flow and efficient particle removal, ensures rapid particle removal in one-pass and recirculating applications. It eliminates the trace levels of ions and improves the quality of ultrapure water.
  • Decreases metal contamination on water and reduces wastes, removes particle greater than 99,9% efficiency at stated retention rating
  • Provides excellent chemical compatibility, long lifetime, reduces changeout and system startup times
Membrane: Patented UPE; Supports, core, sleeve and endcaps: HDPE.
Maximum operating temperature: 60 °C
Maximum forward/reverse differential pressure: 3,5 bar, 50 psid at 25 °C
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité