GateKeeper® GPU 5000 Gas Purifiers
Fournisseur: ENTEGRIS
GPUS5000FEXR02
ENTEGPUS5000FEXR02EA 0 EUR InStock
ENTEGPUS5000FEXR02
GateKeeper® GPU 5000 Gas Purifiers
Filtres Filtres à gaz
GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime and final purity. Leveraging its expertise in material science, Entegris has developed a new generation of purification products that deliver unsurpassed value and performance.
  • Purity control to ppt levels
  • Low pressure drop across the purifier
  • Regenerable
  • Long lifetime
  • Use ambient temperature purification – no heat or power required to operate
  • Insensitive to orientation (horizontal/vertical)
The model 5000 is designed for peak flow rates up to 500 slpm.
Certifications: CE certified to the Pressure Equipment Directive (PED) and manufactured in an ISO® 9001:2000 facility.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité