Planarcore® PVA Brush
Fournisseur: ENTEGRIS INC
PVP0L0TR4
ENTEPVP0L0TR4EA 0 EUR InStock
ENTEPVP0L0TR4
Planarcore® PVA Brush
Brosses
Planarcore® PVA Brush is designed to deliver superior performance and wafer-to-wafer uniformity in post CMP wafer cleaning applications.
  • High-Purity PVA
  • Molded through the core construction
  • Eliminates alignment and gapping problems, increasing system throughput
  • Close molded technology
  • Reduces downtime on tool
  • Decreases defectivity on wafer
The unique molded-through-the-core technology provides absolute adhesion of the PVA to the brush core, unlike standard PVA products that are merely friction fitted to the core.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité