Savana® PTFE Disposable Filters
Fournisseur: ENTEGRIS INC
SVEADS2N1
ENTESVEADS2N1EA 0 EUR InStock
ENTESVEADS2N1
Savana® PTFE Disposable Filters
Filtres Filtres à membrane
Savana® PTFE disposable filters are constructed of PTFE pleated membranes with polypropylene supports and housing to provide fine durability and cleanliness for filtration of process fluids used in the electronics and various industries.
  • The hydrophobic version can be used in IPA or weak solvent applications in flat panel display or hard disk industries
  • Pleated PTFE membrane with polypropylene support and housing
  • Better particle removal performance than polypropylene media
  • Excellent chemical compatibility
  • Suitable for various formats and flow requirements
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité