Savana® Semiconductor Lab Disc Disposable Filter
Fournisseur: ENTEGRIS INC
SVL13D500QQA25
ENTESVL13D500QQA25EA 361 EUR InStock
ENTESVL13D500QQA25
Savana® Semiconductor Lab Disc Disposable Filter
Filtres Disques filtrants
Savana® Lab Disc disposable filters have been specially designed for simple, quick and efficient filtration of liquids used in laboratory, pilot and small-scale applications.
  • Inlet fitting: BD Luer-lok female
  • Outlet fitting: BD Luer-lok female
  • Size: 19 mm
Eight different media options with fitting options can be placed in an all-polypropylene or polyethylene shell construction for excellent chemical compatibility. The small, compact design of the filter capsule also reduces hold-up volume and exposure to hazardous chemicals. No adhesives, binders or surfactants are used in the manufacturing process. The unit is thermally sealed to ensure integrity.
Conditionnement: Pack of 25.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité