PGE1 ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900005
ENZOADI900005EA 536.8 EUR InStock
ENZOADI900005
PGE1 ELISA kit
Tests Tests ELISA
For the quantitative determination of PGE1 in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include simulated interstitial lung fluid.
The PGE1 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405nm. Assay up to 38 samples in duplicate per kit with excellent detection sensitivity.
Informations de commande: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-274 (5x96 wells)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité