Progesterone ELISA Kit
Fournisseur: ENZO LIFE SCIENCES
ADI900011
ENZOADI900011EA 397.1 EUR InStock
ENZOADI900011
Progesterone ELISA Kit
Tests Tests ELISA
For the quantitative determination of progesterone in culture supernatants, serum, and saliva from any species. Cited sample type includes plasma.
The Progesterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in <3 hours. Absorbance is read at 405 nm. This non-radioactive enzyme immunoassay is safe and reliable with ready-to-use liquid color-coded reagents to reduce errors.
Informations de commande: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-280 (5x96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité