cGMP ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900013
ENZOADI900013EA 502.7 EUR InStock
ENZOADI900013
cGMP ELISA kit
Tests Tests ELISA
Most sensitive (0,088pmol/m) colorimetric kit for extracellular cGMP quantification.
For the quantitative determination of cGMP in culture supernatants, plasma, serum, saliva, and urine from any species.
This is a colorimetric competitive immunoassay kit for the quantitative determination of extracellular cGMP. Sensitivity is increased by acetylation (reagents included). Absorbance is read at 405 nm.The ready-to-use liquid color-coded reagents provided reduce error.
Informations de commande: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 2, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine. Alternative size available: 89141-282 (5x96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité