Substance P ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900018A
ENZOADI900018AEA 540.1 EUR InStock
ENZOADI900018A
Substance P ELISA kit
Tests Tests ELISA
Highly sensitive Substance P ELISA kit enabling detection of Substance P in plasma, serum, saliva, urine, and supernatants in just 3 hours.
  • Highly sensitive assay, measuring as little as 5.3 pg/ml of Substance P
  • Widely cited in numerous peer reviewed publications
  • High throughput fomat, assay up to 38 samples in duplicate per 96 well kit
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
The Substance P EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit utilized low sample volume and provides the ability to assay up to 38 samples in duplicate per 96 well kit.
Informations de commande: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Attention: RUO - Research Use Only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité