Leptin (mouse), ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900019A
ENZOADI900019AEA 580.8 EUR InStock
ENZOADI900019A
Leptin (mouse), ELISA kit
Tests Tests ELISA
For the quantitative determination of mouse leptin in culture supernatants, plasma, and serum. Cited sample type includes tissue.
The Leptin (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. The broad dynamic range is suitable for many normal and pathological samples with sensitive quantification of < 26pg/ml mouse leptin.
Informations de commande: Contains Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 28, TMB Substrate, Stop solution 2, Wash buffer concentrate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité