Endothelin-1 ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900020A
ENZOADI900020AEA 785 EUR InStock
ENZOADI900020A
Endothelin-1 ELISA kit
Tests Tests ELISA
The Endothelin-1 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours. This ultra-sensitive kit lets you quantify as little as 0.41 pg/mL ET-1 in a variety of biological matrices with the option of overnight or a 1 hour primary incubation.
  • Highly sensitive measurement
  • Higher throughput format
  • Flexible option of overnight or 1 hour primary incubation
  • Suitable for a variety of sample matrices
For the quantitative determination of ET-1 in culture supernatants, plasma, and serum of human, mouse, rat, cow, dog, pig, and rabbit origin.
Information de livraison: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité