12(S)-HETE ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900050
ENZOADI900050EA 613.8 EUR InStock
ENZOADI900050
12(S)-HETE ELISA kit
Tests Tests ELISA
Sensitive ELISA kit enabling detection of 12(S)-HETE in culture supernatants and plasma from any species.
For the quantitative determination of 12(S)-HETE in culture supernatants and plasma from any species. Cited sample types include serum, renal interstitial fluid and tissue.
The 12(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Fast, accurate, and reliable results are obtained with ready-to-use liquid color-coded reagents that reduce error.
Informations de commande: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-300 (5x96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité