Complement C3a des Arg (human), ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900058
ENZOADI900058EA 776 EUR InStock
ENZOADI900058
Complement C3a des Arg (human), ELISA kit
Tests Tests ELISA
For the quantitative determination of Complement C3a des arg in human EDTA plasma.
The Complement C3a des Arg (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit comes ready-to-use with sample preparation reagents included. The Complement C3a des Arg (human), EIA kit is specific, offering low cross-reactivity to structurally related molecules, and sensitive with quantification of as little as 120pg/ml Complement C3a des Arg.
Informations de commande: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Complement reagent A, Complement reagent B.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité