[pThr180/Tyr182]p38 ELISA Kit
Fournisseur: ENZO LIFE SCIENCES
ADI900101
ENZOADI900101EA 875.6 EUR InStock
ENZOADI900101
[pThr180/Tyr182]p38 ELISA Kit
Tests Tests ELISA
For the quantitative determination of [pThr180/Tyr182]p38 in cell lysates of human and mouse origin.
The [pThr180/Tyr182]p38 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. Use less precious sample per test compared to Western blot with higher throughput sample analysis; measure up to 40 samples in duplicate per kit.
Informations de commande: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité