β-Catenin ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900135
ENZOADI900135EA 806.3 EUR InStock
ENZOADI900135
β-Catenin ELISA kit
Tests Tests ELISA
For the quantitative determination of β-Catenin in cell lysates of human, mouse, and rat origin.
The β-Catenin EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours that measures pan β-Catenin. Absorbance is read at 450 nm. This kit has been validated in-house with human, mouse, and rat cell lysates and correlated Western blot. The β-Catenin EIA kit detects as little 27 pg/ml β-catenin surpassing semi-quantitative Western blot analysis and provides simple liquid color-coded reagents that reduce laboratory error.
Informations de commande: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, RIPA Cell lysis buffer 2, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité