TGF-β1 ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900155
ENZOADI900155EA 691.9 EUR InStock
ENZOADI900155
TGF-β1 ELISA kit
Tests Tests ELISA
For the quantitative determination of TGF-β1 in culture supernatants, plasma, and serum of human, mouse, rat, and cow origin.
The TGF-β1 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450nm. This kit provides ultra-sensitive measurement of as little as 3,3pg/ml TGF-β1 and has been fully validated in activated serum, plasma, and culture supernatants. The ready-to-use liquid color-coded reagents reduce error.
Informations de commande: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Assay buffer 30, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité