Serotonin ELISA Kit
Fournisseur: ENZO LIFE SCIENCES
ADI900175
ENZOADI900175EA 719.4 EUR InStock
ENZOADI900175
Serotonin ELISA Kit
Tests Tests ELISA
The Serotonin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours.
  • Highly sensitive measurement of serotonin
  • No acylation required for standards and samples
  • Validated in multiple complex sample matrices
  • High throughput format with results
  • Fully quantitative results that surpass semi-quantitiave Western blot analysis
For the quantitative determination of Serotonin in platelets, plasma, serum, and urine from any species.
Information de livraison: Supplied with GxR IgG microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité