IL-33 (human) ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900201
ENZOADI900201EA 685.3 EUR InStock
ENZOADI900201
IL-33 (human) ELISA kit
Tests Tests ELISA
IL-33 human ELISA Kit is a colorimetric immunometric immunoassay kit for the quantitative determination of human IL-33 in cell lysates, culture supernatants, serum, plasma and synovial fluid samples.
  • Sensitive - reliably measure as little as 1,7 pg/ml of IL-33
  • Low dynamic range provides sensitive measurements
  • Specific - low cross reactivity to other interleukins
  • Non-radioactive - enzymatic assay format is safe and reliable
  • Easy to use - liquid colour coded reagents reduce errors
Information de livraison: Kit contains microtiter plate, conjugate, antibody, assay buffer 13, wash buffer concentrate, standard, TMB substrate, stop solution 2, xtraction reagent concentrate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité