IL-12p70 (human) ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI900202
ENZOADI900202EA 734 EUR InStock
ENZOADI900202
IL-12p70 (human) ELISA kit
Tests Tests ELISA
The IL-12p70 (human) ELISA Kit is a colorimetric immunometric immunoassay kit for the quantitative determination of human IL-12p70 in culture supernatants, plasma, and serum samples.
  • Specific - no cross reactivity to p40 subunit
  • Rapid - results within 3 hours
  • Sensitive - reliably measure as little as 0,9 pg/ml of IL-12p70
  • Non radioactive - enzymatic assay format is safe and reliable
  • Easy to use liquid colour coded reagents reduce errors
  • Ready to use reagents and pre-coated plates reduce errors
Information de livraison: Kit contains microtiter plate, conjugate, antibody, assay buffer 28, wash buffer concentrate, standard, TMB substrate, and stop solution 2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité