Angiotensin II ELISA Kits
Fournisseur: ENZO LIFE SCIENCES
ADI900204
ENZOADI900204EA 697.4 EUR InStock
ENZOADI900204
Angiotensin II ELISA Kits
Tests Tests ELISA
The Angiotensin II ELISA Kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent Angiotensin II in plasma and serum samples.
  • Results in 3,5 hours
  • Highly sensitive kit detecting as little as 4,6 pg/ml
  • Highly specific with low cross-reactivity to similar peptides
  • Fully quantitative results that surpass semi-quantitative western blot analysis
  • Easy to use liquid colour coded reagents reduce errors
  • Ready to use reagents and pre-coated plates reduce errors and save your time
Cross reactivity: Angiotensin II (100%), Angiotensin A (100%), Angiotensin III (100%), Angiotensin IV (100%), Angiotensin I (0,319%), Angiotensin (1-9) (0,103%), LVV-hemorphin 7 (0,067%), Angiotensin (1-7) (0,053%), Bradykinin (0,022%).
Information de livraison: Kit contains microtiter plate, conjugate, antibody, Assay Buffer 16, wash buffer concentrate, standard, TMB substrate, Stop Solution 2, and Streptavidin-HRP.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité