ELISA kits, LVV hemorphin 7
Fournisseur: ENZO LIFE SCIENCES
ADI900205
ENZOADI900205EA 550 EUR InStock
ENZOADI900205
ELISA kits, LVV hemorphin 7
Tests Tests ELISA
The LVV Hemorphin 7 ELISA Kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent LVV Hemorphin 7 in serum and tissue homogenate samples.
  • Sensitive - reliably measure as little as 6,1 pg/ml of LVV Hemorphin 7
  • Rapid - results in just 3 hours from up to 40 samples in duplicate
  • Specific - low cross reactivity to similar peptides
  • Easy to use liquid colour coded reagents reduce errors
For the quantitative determination of species independent LVV Hemorphin 7 in serum and tissue homogenate samples.
The LVV Hemorphin 7 EIA kit is a colorimetric, competitive immunoassay kit with results in just 3 hours.
Informations de commande: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Information de livraison: The kit contains microtiter plate, conjugate, antibody, assay buffer 16, wash buffer concentrate, standard, TMB substrate, stop solution 2, and Streptavidin-HRP.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité