Survival Motor Neuron (SMN) ELISA Kits
Fournisseur: ENZO LIFE SCIENCES
ADI900209
ENZOADI900209EA 762.3 EUR InStock
ENZOADI900209
Survival Motor Neuron (SMN) ELISA Kits
Tests Tests ELISA
The SMN ELISA Kit is a colorimetric immunometric immunoassay kit for the quantitative determination of human and mouse SMN in cell lysate samples.
  • Highly sensitive measurement, detecting as little as 50 pg/ml of SMN
  • High throughput format with results in <3 hours for up to 39 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
  • Convenient - ready to use liquid colour coded reagents
  • Pre-coated plate saves time and minimise errors
Information de livraison: The kit contains microtiter plate, conjugate, antibody, assay buffer 13, wash buffer concentrate, standard, TMB substrate, stop solution 2, and extraction reagent 4.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité