KIM-1 (human) ELISA kit, grow cells
Fournisseur: ENZO LIFE SCIENCES
ADI9002260001
ENZOADI9002260001EA 578.6 EUR InStock
ENZOADI9002260001
KIM-1 (human) ELISA kit, grow cells
Tests Tests ELISA
Ultra sensitive (1,2 pg/ml) ELISA kit, enabling reduced input sample and matrix interference, for the quantification of KIM-1, an early biomarker for kidney injury or disease.
  • Sensitive: Reliably measure as little as 1,2 pg/ml of KIM-1
  • High Throughput: Results in less than two hours from up to 38 samples in duplicate
  • Specific: Low cross-reactivity to similar proteins
  • Quantitative: Fully quantitative results for both normal and disease state samples that surpass semi-quantitative Western blot analysis
For the quantitative determination of human KIM-1 in urine.
The KIM-1 (human) ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of human KIM-1 in urine with results in less than two hours.
Informations de commande: Contains Microtiter Plate, Assay Buffer, Standard, Antibody-conjugate, Streptavidin-HRP, TMB Substrate, Stop Solution, Wash Buffer Concentrate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité