17β-Estradiol ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI901008
ENZOADI901008EA 1507 EUR InStock
ENZOADI901008
17β-Estradiol ELISA kit
Tests Tests ELISA
Highly sensitive ELISA, measuring as little as 28,5 pg/ml of 17β-Estradiol in < 3 hours.
For the quantitative determination of 17β-Estradiol in culture supernatants and saliva from any species. For serum and plasma use 89141-346.
The 17β-Estradiol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This kit offers specific and save non-radioactive quantification of estradiol with ready-to-use liquid color-coded reagents to reduce error.
Informations de commande: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-060 (1×96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité