Substance P ELISA Kit
Fournisseur: ENZO LIFE SCIENCES
ADI901018A
ENZOADI901018AEA 2176.9 EUR InStock
ENZOADI901018A
Substance P ELISA Kit
Tests Tests ELISA
Highly sensitive substance P ELISA kit enabling detection of substance P in plasma, serum, saliva, urine, and supernatants in just three hours.
  • Highly sensitive assay, measuring as little as 5,3 pg/ml of substance P
  • Widely cited in numerous peer reviewed publications
  • High throughput fomat, assay up to 38 samples in duplicate per 96 well kit
  • Fully quantitative results that surpass semi-quantitative western blot analysis
  • Save with bulk kit package
  • Species independent
  • Wavelength: 405 nm
The Substance P EIA kit is a colorimetric competitive enzyme immunoassay kit with results in three hours. Absorbance is read at 405 nm. This kit utilised low sample volume and provides the ability to assay up to 38 samples in duplicate per 96 well kit.
Substance P is an undecapeptide hormone (RPKPQQFFGLM) that mediates a number of biological activities including nausea, anxiety, pain, and vasodilation. It is a member of the tachykinin family of neuropeptides, and is widely distributed in the peripheral and central nervous systems of vertebrates.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité