Testosterone ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI901065
ENZOADI901065EA 1805.1 EUR InStock
ENZOADI901065
Testosterone ELISA kit
Tests Tests ELISA
For the quantitative determination of testosterone in culture supernatants, plasma, serum, and saliva from any species. Not suitable for human serum and plasma. For those samples use product 89141-348. Cited sample type includes tissue and feces.
The Testosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error. The reproducible results lot-after-lot provide excellent consistency, ideal for longitudinal studies.
Informations de commande: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-112 (1x96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité