ADI901066A
ENZOADI901066AEA
2174.7
EUR
InStock
ENZOADI901066A
Direct cAMP ELISA Kit
Tests
Tests ELISA
Most sensitive (0.006 pmol/ml) colorimetric kit for intracellular cAMP quantification.
- Highly sensitive ELISA with an optional acetylation protocol increasing sensitivity >10-fold
- High troughput format with results in <3 hours for up to 39 samples in duplicate
- Easy-to-use pre-coated plate and liquid color-coded reagents minimizes errors
- Simple, efficient and well-established sample handling protocols
- Highly reliable with consistent lot-to-lot performance
This is a colorimetric competitive immunoassay kit for the quantitative determination of intracellular cAMP in cells or tissues lysed in 0,1 M HCl, which is used to stop endogenous phosphodiesterase activity and stabilize the released cyclic AMP. The 0,1 M HCl treated samples are then analyzed directly in a microtiter plate without extraction, drying and reconstitution.
Sensitivity is increased >10-fold by acetylation (reagents included). Absorbance is read at 405 nm. The large signal-to-background ratio offers superior sensitivity compared to competitors. This kit lets you easily assess adenylyl cyclase activation by GPCRs.
Cyclic AMP (cAMP) is one of the most important "second messengers" involved as a modulator of physiological processes. A number of hormones are known to activate cAMP through the action of the enzyme Adenylate cyclase which converts ATP to cAMP. cAMP has been shown to be involved in the cardiovascular and nervous systems, immune mechanisms, cell growth and differentiation, and general metabolism.
Shipping conditions: Blue ice