PGF2α ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI901069
ENZOADI901069EA 1955.8 EUR InStock
ENZOADI901069
PGF2α ELISA kit
Tests Tests ELISA
For the quantitative determination of PGF2α in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include cell lysate and whole blood.
The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors.
Informations de commande: Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-120 (1x96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité