Cysteinyl leukotriene ELISA kit
Fournisseur: ENZO LIFE SCIENCES
ADI901070
ENZOADI901070EA 2210 EUR InStock
ENZOADI901070
Cysteinyl leukotriene ELISA kit
Tests Tests ELISA
Highly sensitive ELISA kit enabling detection of LTC4/D4/E4 in 4 hours.
For the quantitative determination of Cysteinyl Leukotrienes (LTC4/D4/E4) in culture supernatants and urine from any species.
The Cysteinyl Leukotriene EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Safe non-radioactive measurement of LTC4/D4/E4. Ready-to-use liquid color-coded reagents reduce errors.
Informations de commande: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-122 (1x96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité