Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-AGTR1 Rabbit Polyclonal Antibody (DyLight® 682)

Fournisseur: ENZO LIFE SCIENCES
ADI905813100
ENZOADI905813100EA 471.9 EUR InStock
ENZOADI905813100
Anti-AGTR1 Rabbit Polyclonal Antibody (DyLight® 682)
Anticorps
The octapeptide circulatory hormone Angiotensin II is a critical regulator of cardiovascular homeostasis formed in blood, with dual function in tissues of the brain, kidney, heart, and vasculature as an autocrine/paracrine growth hormone. The actions of angiotensin II are mediated by type one (AT1) and type two (AT2) angiotensin II receptors, members of the seven-transmembrane Gprotein coupled receptor (GPCR) family. AT1 receptors couple predominantly with Gq/11 alpha subunits, and signal through phospholipases C, D, and A2, inositol phosphates, and calcium channels, as well as a variety of serine/threonine and tyrosine kinases. AT1 expression is greatest in vascular smooth muscle, liver, kidney, adrenal cortex, brain, and lung, and is primarily responsible for the physiological effects of angiotensin II, including regulation of arterial blood pressure, electrolyte and water balance, thirst, hormone secretion, and renal function. The AT2 receptor displays approximately 30% identity to the AT1 receptor, and is highly expressed during embryonic development. AT2 expression in adult tissue is limited to the adrenal medulla, brain, and reproductive tissues, but is upregulated in injured tissue following myocardial infarction, vascular injury, and wound healing. AT2 receptor signaling is not dependent on G-protein coupling or receptor phosphorylation, although coupling to Gia2 and Gia3 alpha subunits occurs in some tissues. AT2 activation can stimulate tyrosine and serine/threonine phosphatases through activation of nitric oxide and cGMP, and appears to counteract the stimulatory effects of AT1 receptor and growth factor receptor signaling.

Recognizes human, mouse and rat angiotensin II receptor AT1. Predicted MW of ~41kDa. Higher MW species (~55kDa) and intermediates also observed by Western blot which reflect post-translational modification and dimerization of the receptor. Predicted species reactivity based on epitope sequence homology: chimpanzee, bovine, rhesus macaque, sheep, dog, guinea pig, rabbit, horse, wild boar, gerbil.

For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Informations de commande: Manufactured by Proteimax.

Type: Primary
Antigen: AGTR1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 682
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité