Catalase fluorometric detection kit
Fournisseur: ENZO LIFE SCIENCES
ADI907027
ENZOADI907027EA 533 EUR InStock
ENZOADI907027
Catalase fluorometric detection kit
Tests Tests cellulaires
The Catalase fluorometric detection kit provides a higher throughput sensitive method for measuring catalase activity in just one hour. This is a homogeneous mix-and-read assay that enables you to monitor multiple time points for a kinetic read. Positive control is provided.
  • Easily adaptable to HTS applications
  • Sensitive fluorescent detection system
For the measurement of catalase in cell lysates from any species.
Information de livraison: Reaction buffer concentrate, Detection Reagent, Horseradish peroxidase concentrate, Hydrogen peroxide, Catalase enzyme
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité