Myeloperoxidase fluorometric detection kit
Fournisseur: ENZO LIFE SCIENCES
ADI907029
ENZOADI907029EA 924 EUR InStock
ENZOADI907029
Myeloperoxidase fluorometric detection kit
Tests Tests cellulaires
The Myeloperoxidase fluorometric detection kit is a one-step no wash homogeneous activity assay that is adaptable to HTS applications. This kit offers highly sensitive fluorescent detection and the ability to monitor multiple time points to follow the kinetics of MPO activity.
  • Fast homogenous assay
  • Suitable for High Throughput Screening
  • Monitor multiple time points to follow kinetics
  • Results in 30 to 60 minutes
For the measurement of Myeloperoxidase activity in cells and tissue from any species.
Information de livraison: Assay buffer concentrate, Detection Reagent, Hydrogen peroxide, Myeloperoxidase enzyme
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité