Human recombinant irisin (active)
Fournisseur: ENZO LIFE SCIENCES
ADI9083070010
ENZOADI9083070010EA 515.9 EUR InStock
ENZOADI9083070010
Human recombinant irisin (active)
Protéines et Peptides
Irisin is a hormone that is involved in converting white adipose tissue into brown adipose tissue after physical exercise.
  • Multifunctional protein validated for activity assay, ELISA, SDS-PAGE and Western blot
  • Important regulatory protein for metabolism and neurogenesis
  • Produced in CHO cells, comprises the majority of the extracellular domain of FNDC5
  • Plays an important role in energy regulation and furthermore may lead to improvements in glucose regulation, insulin sensitivity, hypertension, and obesity
Irisin is formed after the membrane bound protein fibronectin type III domain containing protein 5 (FNDC5) is cleaved resulting in a secreted protein. FNDC5 is found in muscle tissue and is activated during physical exertion via the protein PGC1a. Soluble Irisin has also been shown to up regulate UCP-1 gene expression in mice. UCP-1 is a mitochondrial transporter protein and a marker of brown adipose tissue conversion.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité