PGE2 FPIA kit
Fournisseur: ENZO LIFE SCIENCES
ADI920001
ENZOADI920001EA 378.4 EUR InStock
ENZOADI920001
PGE2 FPIA kit
Tests Tests ELISA
For the quantitative determination of PGE2 in culture supernatants from any species.
The PGE2 FPIA kit is a homogeneous mix-and-read fluorescence polarization enzyme immunoassay kit with. Results are obtained within just 30 minutes and the generated signal is stable for up to 20 hours. The PGE2 FPIA kit is ideal for high throughput screening (HTS) applications. A convenient homogeneous mix-and-read assay that is amendable to miniaturization and automation, this kit has a robust Z factor >0.8.
Informations de commande: Contains Conjugate concentrate, Antibody, Assay buffer concentrate, Standard.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité