Anti-Fas Mouse Monoclonal Antibody [clone:ZB4]
Fournisseur: ENZO LIFE SCIENCES
ADIAAM227E
ENZOADIAAM227EEA 437 EUR InStock
ENZOADIAAM227E
Anti-Fas Mouse Monoclonal Antibody [clone:ZB4]
Anticorps
Fas/APO-1/CD95 is type I-membrane glycoprotein that is a member of the Tumor necrosis factor receptor (TNF-R) family, which includes TNF-R1, TNF-R2, CD27, CD30, and CD40.
  • Dilution range: 5 to 20 µg/ml for Flow Cytometry, 5 µg/ml, colorimetric for Western blot
  • Detects a band of ~45 kDa by Western blot
Binding of Fas-ligand (FasL) to Fas or crosslinking of Fas by anti-Fas monoclonal antibodies leads to apoptosis of Fas expressing cells. Apoptosis induced by Fas-Fas ligand interaction is inhibited by a number of cytosolic factors such as the ICE/CED3More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité