PKA kinase activity kit
Fournisseur: ENZO LIFE SCIENCES
ADIEKS390A
ENZOADIEKS390AEA 731.5 EUR InStock
ENZOADIEKS390A
PKA kinase activity kit
Tests Tests enzymatiques
The PKA Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the quantitative determination of PKA kinase activity in partially purified, purified, or crude enzyme preparations from any species.
Informations de commande: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité