p70 S6K activity kit
Fournisseur: ENZO LIFE SCIENCES
ADIEKS470
ENZOADIEKS470EA 855.8 EUR InStock
ENZOADIEKS470
p70 S6K activity kit
Tests Tests cellulaires
The p70 S6K activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the measurement of p70 S6K activity in partially purified, purified, or crude enzyme preparations from any species.
Informations de commande: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité