HSP27 (human), ELISA Kit
Fournisseur: ENZO LIFE SCIENCES
ADIEKS500
ENZOADIEKS500EA 918.5 EUR InStock
ENZOADIEKS500
HSP27 (human), ELISA Kit
Tests Tests ELISA
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of human HSP27 in cell lysates, plasma, serum, and tissue.
The HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Highly sensitive quantitative determination of Hsp27 independent of phosphorylation state.
Informations de commande: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité