Anti-HSP70 IgG/A/M (human), ELISA Kit
Fournisseur: ENZO LIFE SCIENCES
ADIEKS750
ENZOADIEKS750EA 939 EUR InStock
ENZOADIEKS750
Anti-HSP70 IgG/A/M (human), ELISA Kit
Tests Tests ELISA
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of anti-HSP70 IgG/A/M autoantibodies in human serum.
This assay allows for reproducible, accurate and precise determination of IgG , IgA and IgM antibodies (total) to human Hsp70 in serum. The ELISA kit uses recombinant human Hsp70 bound to the wells of the plate to bind anti-human Hsp60 antibodies present in human serum. The captured anti-human Hsp70 antibodies are detected with a HRP conjugated goat polyclonal antibody specific for human IgG, IgA and IgM molecules. The assay is developed with tetramethylbenzidine substrate pro ducing a blue color in proportion to the amount of captured anti -human Hsp70 antibodies. The color development is stopped with acid stop solution which converts the endpoint color to yellow. The intensity of the color is measured in a microplate reader at 450 nm.
Informations de commande: Contains Microtiter plate, Standard, Sample diluent 2, Wash buffer concentrate, HRP Conjugate, TMB Substrate, Stop solution 2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité