HSP90α (human), ELISA Kit
Fournisseur: ENZO LIFE SCIENCES
ADIEKS895
ENZOADIEKS895EA 1098.9 EUR InStock
ENZOADIEKS895
HSP90α (human), ELISA Kit
Tests Tests ELISA
Most sensitive, rapid ELISA kit for highly specific, quantitative detection of human HSP90α.
For the quantitative determination of HSP90α in cell lysates, serum, and tissue of human origin.
The HSP90α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Informations de commande: Contains Microtiter plate, 5X Extraction reagent 2, Standard, Sample diluent, Wash buffer concentrate, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité