Anti-BVR Rabbit Polyclonal Antibody
Fournisseur: ENZO LIFE SCIENCES
ADIOSA400E
ENZOADIOSA400EEA 517 EUR InStock
ENZOADIOSA400E
Anti-BVR Rabbit Polyclonal Antibody
Anticorps
Cleavage of heme b (Fe-protoporphyrin IX) at the a-methene carbon bridge to form the open tetrapyrrole, biliverdin IXa and carbon monoxide (CO) is catalyzed by heme oxygenase (HO) isozymes HO-1 and HO-2 (heme hydrogen-donor: oxygen oxidoreductase; EC 1.14.99.3). In mammalian species, biliverdin reductase (BVR; bilirubin: NAD(P)+ oxidoreductase; EC 1.3.1.24) converts the open tetrapyrrole to bilirubin. This pathway represents the only efficient way of making bilirubin and thereby deterring activation of oxygen by the heme molecule. HO-1 belongs to the heat shock protein family (Hsp32), whileMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité