Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-HSP91 Rat Monoclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
ADISPA845F
ENZOADISPA845FEA 598 EUR InStock
ENZOADISPA845F
Anti-HSP91 Rat Monoclonal Antibody
Anticorps
The 90 kDa molecular chaperone family includes 90 kDa heat shock protein Hsp90 and 94 kDa glucose-regulated protein grp94, both major molecular chaperones of the cytosol and the endoplasmic reticulum. Mammalian cells contain isoforms Hsp90α and Hsp90β, encoded by separate genes. The amino acid sequences of human and yeast Hsp90-alpha are 85% and 90% homologous to those of Hsp90β , respectively. All known members of the Hsp90 protein family are highly conserved, especially in the N-terminal and C-terminal regions containing independent chaperone sites with different substrate specificity. These ubiquitous and highly conserved proteins account for 1 to 2% of all cellular proteins in most cells. Hsp90 functions as part of the cell’s powerful network of chaperones to fight the deleterious consequences of protein unfolding caused by non-physiological conditions. In the absence of stress, however, Hsp90 provides a necessary component of such fundamental cellular processes as hormone signaling and cell cycle control. In this context, researchers identified key regulatory proteins as substrates of Hsp90, including steroid receptors, cell cycle kinases involved in signal transduction, and p53. Hsp90 may act as a capacitor for morphological evolution by buffering widespread variation, potentially affecting morphogenic pathways. When temperature and other stress factors compromise Drosophila Hsp90 buffering, cryptic variant expression occurs, and selection can lead to the continued expression of these traits even after Hsp90 function is restored.

  • Dilution range: Western Blot (1:500, colorimetric)
  • Detects a band of ~90kDa by Western blot

Recognizes human, mouse, rat, beluga, bovine, dog, chicken, fish, guinea pig, hamster, monkey, rabbit, sheep, and water mold HSP90. Detects a band of ~90kDa by Western blot.

Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone: 2D12
Conjugation:
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Beluga, Bovine, Chicken, Dog, Fish, Guinea pig, Hamster, Human, Monkey, Mouse, Rabbit, Rat, Sheep, Water mold

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité