MALP-2 (BULK)
Fournisseur: ENZO LIFE SCIENCES
ALX162034C500
ENZOALX162034C500EA 2256.1 EUR InStock
ENZOALX162034C500
MALP-2 (BULK)
Protéines et Peptides
TLR activator
MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. For description of the stereochemistry of MALP-2. MALP-2 signalling, unlike that of LPS, is not transduced via TLR4, but is induced via TLR2 and TLR6 signalling. For more information about MALP-2 see www.malp-research.de. BULK size (500 µg) of biologically active, pyrogen-free and sterile MALP-2 for in vitro and animal in vivo studies.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité