Human recombinant Control:Fc fusion protein
Fournisseur: ENZO LIFE SCIENCES
ALX203004C050
ENZOALX203004C050EA 585.2 EUR InStock
ENZOALX203004C050
Human recombinant Control:Fc fusion protein
Protéines et Peptides
Negative control for Fc chimeric proteins
Produced in HEK 293 cells. The extracellular domain of mutated hThy-1, hThy-1 (RLE) (aa 20-130), is fused to the Fc portion of human IgG1. The integrin binding sequence RLD of hThy-1 has been changed to RLE abolishing the binding of integrins.
Negative control for Fc chimeric proteins. Does not bind to a series of ligands in an ELISA assay. Does not inhibit binding between the thymoma cell line EL-4 expressing β3-integrin and astrocytes expressing high level of Thy-1 in mice.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité