Synonyms:
2-(p-(2,2-Dichlorocyclopropyl)phenoxy)-2-methylpropionic acid, 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropionic Acid, 2-(4-(2,2-Dichlorocyclopropyl)phenoxy)-2-methylpropanoic acid
ALX270475M025
ENZOALX270475M025EA
84.7
EUR
InStock
ENZOALX270475M025
Ciprofibrate ≥98% (par HPLC)
Ciprofibrate
PPARα activator.
Hypolipidemic compound. PPARα (peroxisome proliferator-activated receptor α) agonist.
Soluble in DMSO.