Microcystis aeruginosa Microcystin HILR
Fournisseur: ENZO LIFE SCIENCES
ALX350177C025
ENZOALX350177C025EA 275 EUR InStock
ENZOALX350177C025
Microcystis aeruginosa Microcystin HILR
Protéines et Peptides
Microcystins are a group of cyclic heptapeptide hepatotoxins produced by a number of cyanobacterial genera. The most notable of which, and namesake, is the widespread genus Microcystis. Structurally, all microcystins consist of the generalized structure cyclo(-D-Ala1-X2-D-MeAsp3-Y4-Adda5-D-Glu6-Mdha7-). X and Y are variable L-amino acids, D-MeAsp is D-erythro-β-methylaspartic acid and Mdha is N-methyldehydroalanine. Adda is the cyanobacteria unique C20 β-amino acid 3-amino-9-methoxy-2,6,8-trimethyl-10-phenyl-deca-4,6-dienoic acid. Substitutions of the variable L-amino acids at positions 2 and 4 give rise to at least 21 known primary microcystin analogs and alterations in the other constituent amino acids result in more than 90 reported mycrocystins to date.
Isolated from Microcystis aeruginosa. Microcystin containing a homoisoleucine (Hil) in position 2 and arginine (R) in position 4. Hepatotoxic.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45a7464eb03d17f72e864bc47fa389a28763de6a&vid=564a5b936a85ca62c6a35cc8a7d97ee3d8e48215&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité