Anabaenopeptin B ≥95% (par HPLC)
Fournisseur: ENZO LIFE SCIENCES
ALX350184C100
ENZOALX350184C100EA 277.2 EUR InStock
ENZOALX350184C100
Anabaenopeptin B ≥95% (par HPLC)
Anabaenopeptin B
Inhibitor of protein phosphatase.
Isolated from Microcystis aeruginosa.
Anabaenopeptins are a highly diverse group of bioactive peptides produced by several genera of cyanobacteria such as Anabaena, Planktothrix, Microcystis and Nodularia. These peptides are commonly detected in cyanobacterial blooms along with the well known microcystins. Unique cyclic peptides where lysine is conserved in position 2 and cyclisation is achieved by linkage of the lysine to the C-terminal amino acid. In addition the side chain of a single amino acid is attached via a ureido bond. Bioactive peptides originally isolated from Anabaena flos-aquae NRC 525-17 where they were shown to be non-toxic in mice but had dose dependant relaxations in rat aortic preparations with endothelium precontracted with 0.1µM norpinephrine.
Anabaenopeptins inhibit protein phosphatase.
Numéro CAS: 161897-74-1
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité